Colourimetric test kit, Phosphatest®, TESTOVAL®
HEYL410593
:
- Cdt:180 Tests
- Désignation:ortho-Phosphate, refill pack, 1 set of reagents
- Seq:02
- Plage de mesure:0,25-0,5-1-2,5-5-10 mg/l
Colour comparison kit for the rapid manual analysis of the phosphate content in water in the concentration range 0 - 10 mg/l.
The individual values can be read by optical colour comparison. By diluting the water sample 1:10 with distilled water, the measuring range can be increased to 10 times the concentration.
: Supplied with colour comparator.