D(+)-Glucose monohydraté
Supplier: Chemicals
Jump to:
About this item
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Melting Pt:83 °C
- Storage Temperature:Température ambiante
- EINECS:200-075-1
3 Options Available Below
Product Details & Documents
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Melting Pt:83 °C
- Storage Temperature:Température ambiante
- EINECS:200-075-1
Product Family Options
Product Information
- Type de conditionnementCdtAvailabilityPrice
- Specifications:MarketsourceMarketsource
This product has been blocked by your organization. Please contact your purchasing department for more information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



