Porous polymer adsorbent, Tenax®
11982.
: Merck
:
- Cdt:10 g
- Désignation:Tenax® porous polymer adsorbent, 10 g
- Mailles:60 – 80 mesh
- Emballage:Bottle
These porous polymers, based on 2,6-diphenyl-p-phenylene oxide, are widely used as an adsorbent in both air collection and purge trap applications.
The unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.
Surface area: ~35 m²/g
Density: ~0,25 g/ml (free fall density)