Slide stainer
Slide stainer and dyer, SS-30, 30
KLINSS-30
Slide stainer designed to be used for Hematoxylin and Eosin (H&E), Papanicolau (PAP) and user-specified staining protocols.
- Bilateral robotic arm: Free choice of the station sequence in a staining protocol
- Staining capacity of up to five racks simultaneously, depending on protocols, load frequency and instrument configuration
- USB connection for programming of staining protocols and data gathering
The Reagent Management System (RMS) controls the use of reagents and helps to achieve a better staining quality.
: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : [email protected]
- Modèle:SS-30
- Nbre de lames:30
- Désignation:Slide stainer and dyer