Z-AAF-CMK
ENZOBMLPI1240005
:
- Cdt:5 mg
- Seq:02
- Nom de la protéine/du peptide:Z-AAF-CMK
- Purity:98%
Negative control for AAF-CMK
Negative control for serine protease inhibitor AAF-CMK which inhibits tripeptidyl peptidase II (TPPII), a giant protease which may substitute for some proteasome functions. Blockage of the N-terminus by the Benzyloxycarbonyl or “Z” group abolishes the activity of the AAF-CMK inhibitor.