AIP
Catalog # ENZOBMLP2110001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Cdt:1 mg
- Seq:02
- Synonyme de protéine:Autocamtide-2-related inhibitory peptide
- Nom de la protéine/du peptide:AIP
Specifications
About this item
CaM kinase inhibitor
AIP (autocamtide-2-related inhibitory peptide) is a highly specific and potent inhibitor of CaM kinase II. This peptide is derived from the CaM kinase II substrate autocamtide-2 (BML-P101) but alanine is substituted for threonine at the 9-position. AIP at 1 µM completely inhibited CaM kinase II but did not affect PKC, PKA, CaMKIV and other brain extract kinases. A cell-permeable, myristoylated version is available (BML-P212).