ContactUsLinkComponent
CA-074
  ENZOALX260017M001
 :  
CA-074
  ENZOALX260017M001
 :  
  • Cdt:
    1 mg
  • Seq:
    02
  • Nom de la protéine/du peptide:
    CA-074
Cathepsin B inhibitor
Potent and specific inhibitor of cathepsin B in vitro and in vivo. Free acid form used in vitro or in isolated enzyme studies but not in whole cells. Can discriminate between cathepsin B and L/S forms.