CA-074
ENZOALX260017M001
:
- Cdt:1 mg
- Seq:02
- Nom de la protéine/du peptide:CA-074
Cathepsin B inhibitor
Potent and specific inhibitor of cathepsin B in vitro and in vivo. Free acid form used in vitro or in isolated enzyme studies but not in whole cells. Can discriminate between cathepsin B and L/S forms.