alphaRsaalphaI
N° de catalogue ENZOENZGEN1091000
Fournisseur: ENZO LIFE SCIENCES
Numéro CAS:
Spécifications
- Cdt:1 Vial
- Seq:02
- Synonyme de protéine:CGH|restriction enzyme|SNP|cloning|endonuclease|digest|digestion|mapping|southern blotting
- Nom de la protéine/du peptide:AlphaRsaAlphaI
- Concentration:RsaI 10000 units/ml. Includes 10X cutting buffer.
- Formulation:Liquid. In 100 mM sodium chloride, containing 10 mM Tris-HCl, pH 7,4, 1 mM DTT, 0,1 mM EDTA, 50% glycerol and 200 µg/ml BSA.After reconstitution, 1X Cutting Buffer: 50 mM potassium acetate, 20 mM Tris-acetate, 10 mM magnesium acetate, 100 µg/ml BSA, pH 7,9, at 25 ℃.
Spécifications
À propos de cet article
Restriction enzyme for molecular biology applications
- High quality enzyme with stringent QC testing
- Includes 10X Cutting Buffer
- BSA premixed into buffers
- Suitable for molecular cloning, restriction site mapping, genotyping, southern blotting, SNP
Produced in an E. coli strain that carries the cloned RsaI gene from Rhodopseudomonas sphaeroides.
RsaI is a restriction enzyme that recognizes GT^AC sites.10X Cutting Buffer is included that contains BSA, which enhances enzyme stability and binds to contaminants in DNA preps.