Hsp27tide
1 mg
N° de catalogue ENZOBMLP2460001
Fournisseur: Enzo Life Sciences
Aller à :
Product Details & Documents
MAPKAPK substrate
This peptide, RRLNRQLSVA, is derived from Hsp27 (aa 80-85) and is a substrate for MAPKAPK2 and MAPKAPK3.
Spécifications
Spécifications
- Cdt:1 mg
- Cat. No.:ENZOBMLP2460001
- Purity:95%
- Nom de la protéine/du peptide:Hsp27tide