ContactUsLinkComponent
ERK1/ERK2 (172-190)
ERK1/ERK2 (172-190)
  ENZOBMLP1240001
 :  
ERK1/ERK2 (172-190)
  ENZOBMLP1240001
 :  
  • Cdt:
    1 mg
  • Seq:
    02
  • Nom de la protéine/du peptide:
    ERK1/ERK2 (172-190)
  • Purity:
    97%
MEK substrate
This sequence, ADPDHDHTGFLTGYVATRTRR, is based on Erk1/Erk2 residues 172-190, containing a MAP kinase kinase phosphorylation site (underlined). N-terminal Arg is added for phosphocellulose binding.