Mouse;Rat obestatin
1 mg
N° de catalogue ENZOALX164005M001
Fournisseur: Enzo Life Sciences
Aller à :
Product Details & Documents
Food intake modulator
Obestatin derives like ghrelin from proghrelin, but exhibits opposite physiological effects. Obestatin decreases food intake and body weight in rodents. Moreover, obestatin also counteracts ghrelin’s effect by decelerating gastric emptying and decreasing intestinal contractility. Obestatin binds to orphan G protein-coupled receptor GPR39. Amidation of the C-terminus seems to be important for obestatin’s activity.
Spécifications
Spécifications
- Cdt:1 mg
- Cat. No.:ENZOALX164005M001
- Formulation:Lyophilised
- Purity:>97% (HPLC)
- Species:Rat|Mouse
- Nom de la protéine/du peptide:Obestatin
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



