ContactUsLinkComponent
EpiXtract® total histone extraction kit (high throughput)
EpiXtract® total histone extraction kit (high throughput)
N° de catalogue ENZOENZ450130002
Fournisseur:  ENZO LIFE SCIENCES
Numéro CAS:  
undefined
EpiXtract® total histone extraction kit (high throughput)
N° de catalogue ENZOENZ450130002
Fournisseur:  ENZO LIFE SCIENCES
Numéro CAS:  
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    EpiXtract® total histone extraction kit (high throughput)
  • Seq:
    02
Spécifications
À propos de cet article
Rapid, reliable, high throughput method to isolate histone proteins in mammalian cells for histone modification applications.
For the extraction of total histone proteins from mammalian cells in a 96 well plate format.
EpiXtract® Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in one hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Informations de commande: Contains Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .