Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Caspase 9
- Symbole de l'antigène:CASP9
- Clonalité:Monoclonal
- Hôte:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Seq:02
- Formulaire:Liquid. In PBS, pH 7,2, containing 0,09% sodium azide and 50% glycerol.
- Synonymes antigène:EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
- Purification:Protein G affinity purified
- Cdt:100 µG
Spécifications
À propos de cet article
The caspases are a family of cysteine proteases that cleave after certain aspartate residues, and are primarily recognized as mediators of apoptosis. caspases are synthesized as inactive zymogens that can be cleaved to form active enzymes following the induction of apoptosis by stress or death receptors. Initiator caspases (e.g. caspase-8 and -10) are activated by dimerization of the zymogen on a dedicated adaptor protein. These activated initiator caspases in-turn cleave downstream effector or executioner caspases (e.g. caspase-3, -6, and -7) in a cascade-like manner, which cleave key cellular proteins that lead to the morphological changes associated with apoptotic cell death.
Type: Primary
Antigen: CASP9
Clonality: Monoclonal
Clone: 32051
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human