Eyewash station
Catalog # LBES8205203
Supplier: LABORATOIRES ESCULAPE
Specifications
- Désignation:Eyewash station, with 200 ml pH neutral solution and 500 ml sterile sodium chloride solution, 230×80×300 mm
- Environmentally Preferable:
- Cdt:1 KIT
Specifications
About this item
This eyewash station contains one 200 ml bottle with a pH neutral solution and one 500 ml bottle with an ophthalmic solution (sterile sodium chloride solution). The pH neutral solution is a sterile phosphate buffer (4,9%) that effectively and rapidly neutralises splashes of alkaline (base) or acidic solutions in the eyes or on the skin and which brings the pH level back to a normal level.
- Neutral pH solution (blue bottle) for neutralising acids and alkalis
- Sterile solution of sodium chloride (green bottle) for cleaning and rehydrating the eye
- With mirror and pictograms
Rinsing time: 7 minutes
Shelf life: 3 years