Platinum/iridium wire
Platinum/iridium wire, Ø 0,8 mm
  631-7116
 :   CHEMPUR
Platinum/iridium wire
  631-7116
 :  CHEMPUR
Platinum/iridium wire
Platinum/iridium wire, Ø 0,8 mm
1 * 1 cm
Platinum/Iridium 90/10 wire.
Platinum wire is used in many applications including gasket or o-ring fabrication, thermocouples, electrical contacts, loops and heater coils.
  • Désignation:
    Platinum/iridium wire, Ø 0,8 mm
  • Seq:
    01