Solid phase extraction disk, ENVI™-18
Solid phase extraction disk, ENVI™-18
Solid phase extraction disk, ENVI™-18, 47 mm
N° de catalogue 57171.
Fournisseur:   Merck
Solid phase extraction disk, ENVI™-18
N° de catalogue 57171.
Fournisseur:  Merck
Solid phase extraction disk, ENVI™-18
Solid phase extraction disk, ENVI™-18
Solid phase extraction disk, ENVI™-18, 47 mm
Prix
273,00 €
Connexion Pour la disponibilité
1 * 24 ST
Product Details & Documents
Solid phase extraction disks provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples.
  • Reversed phase retention mechanism
  • Porous glass fiber membranes embedded with C18 or C8 modified silica particles
Applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides.
Spécifications
  • Ø:
    47 mm
  • Désignation:
    Solid phase extraction disk, ENVI™-18