Ross-Miles apparatus
Catalog # NEUB2749-90-010
Supplier: NEUBERT VOLUME GLASSWAERE
Specifications
- Désignation:Complete Ross-Miles apparatus
- Cdt:1
Specifications
About this item
This Ross-Miles apparatus is used for testing of surfactants.
It is specially necessary during the determination of foaming powder.
Certifications: DIN 53902