Reference standards, surface water - trace metals
Fournisseur: LGC Standards PROMOCHEM
Certificats
À propos de cet article
This reference standard is intended for daily use in conjunction with measurement of elements in surface waters. The water solution contains 45 elements with concentrations traceable to
ultrapure metals or stoichiometrically well-defined substances.
Frequently Bought Together