Standards for water pollution QC check, sample hardness, VHG
PROOVHG-QWSHRD-250
:
- Cdt:250 mL
- Désignation:Water analysis standard for hardness
- Type:Reference standard
This product is intended for use as a quality control check for methods related to hardness measurements includes calcium, magnesium, sodium, calcium hardness and total hardness as CaCO₃.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
: Traceable to NIST SRM.