Standards for water pollution QC check, minerals, VHG
PROOVHG-QWPMIN-500
:
- Cdt:500 mL
- Désignation:Water analysis standard for minerals
- Type:Reference standard
This solution is intended for use as a quality control check for methods related to minerals.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
: Traceable to NIST SRM.