Human serum, multiple sclerosis (MS)
Product Details & Documents
Human serum used for researching the causes, diagnosis, supportive care and treatment of multiple sclerosis (MS). Collected from IRB-consented donors diagnosed with MS.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Spécifications
- Format du sérum:Cryopreserved
- Désignation:Serum, multiple sclerosis (MS)