Afterwash solution
Afterwash solution
Afterwash solution, 200 ml bottle
Catalog # LBES8205212
Afterwash solution
Catalog # LBES8205212
Afterwash solution
Afterwash solution
Afterwash solution, 200 ml bottle
Price
48,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
This afterwash solution quickly restores the natural physiological condition of the eye following decontamination with Diphoterine® or Hexafluorine® solutions.
  • Ergonomically eye-shaped cup ensures a tighter seal around the eye to maximise efficiency during rinsing
  • Helps to restore the physiological state of the cornea more quickly and more smoothly
  • May be used for eye wash in case of non chemical solid particles
Shelf life: 2 years
Specifications
  • Désignation:
    Afterwash solution, 200 ml bottle
  • Seq:
    03