ContactUsLinkComponent
Human recombinant proteoglycan 3 (from cells)
Human recombinant proteoglycan 3 (from cells)
Catalog # PRSI91-673
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant proteoglycan 3 (from cells)
Catalog # PRSI91-673
Supplier:  ProSci Inc.
CAS Number:  
Specifications
  • Cdt:
    50 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Conditions de stockage:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Synonyme de protéine:
    Prepro-MBPH|Prepro-Major Basic Protein Homolog|MBPH|Proteoglycan 3|Eosinophil Major Basic Protein Homolog|PRG3
  • Nom de la protéine/du peptide:
    Proteoglycan 3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    24.6 kD
  • Sequence:
    Leu18-Phe225
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
Specifications
About this item
Proteoglycan 3, also known as Eosinophil major basic protein homolog, Prepro-major basic protein homolog, PRG3 and MBPH, contains one C-type lectin domain. Proteoglycans are a major component of the animal extracellular matrix. PRG3 localizes to the eosinophil secondary granule and is expressed in bone marrow, not detected in placenta. PRG3 has similar cytotoxic and cytostimulatory activities to PRG2/MBP. In vitro, PRG3 can stimulate neutrophil superoxide production and IL8 release, histamine and leukotriene C4 release from basophils.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag