Anti-EIF4E Rabbit Polyclonal Antibody
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
- Taille:100 µg
- Western blot:Yes
- Formulaire:Liquid
- Symbole de l'antigène:EIF4E
- Synonymes antigène:eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
- ImmunoFluorescence:Yes
- Conjugaison:Unconjugated
- Concentration:1 mg/mL
- Réactivité:Human
- Cat. No.:PRSI79-211
- Clonalité:Polyclonal
- Immunogène:eIF4E (Phospho-Ser209) antibody was raised against a peptide sequence around phosphorylation site of serine 209 (S-G-S (p) -T-T) derived from Human eIF4E.
- Hôte:Rabbit
- ImmunoChimie:Yes



