Anti-ACTN2 Rabbit Polyclonal Antibody
Product Details & Documents
ACTN2 Antibody can be used for Immunocytochemistry, Immunofluorescence starting at 1:100 - 1:200, Immunohistochemistry in paraffin embeded tissues starting at 10μg/mL, and Western Blotting starting at 1:500 - 1:3000.
0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal
0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ACTN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10075-212.pdf
Fiche technique
Spécifications
- Taille:50 µl
- Western blot:Yes
- Symbole de l'antigène:ACTN2
- Synonymes antigène:CMD1AA|CMH23
- ImmunoFluorescence:Yes
- Conjugaison:Unconjugated
- Concentration:1 mg/mL
- Réactivité:Human
- Cat. No.:PRSI70-845
- Clonalité:Polyclonal
- Immunogène:Recombinant protein fragment containing a sequence corresponding to a region within amino acids 215 and 500 of actinin, alpha 2.
- Nom de l'antigène:Actinin alpha 2
- Hôte:Rabbit
- ImmunoChimie:Yes
- Tested applications:Immunoaffinity Chromatography
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



