Anti-EIF4E3 Rabbit Polyclonal Antibody
Product Details & Documents
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF4E3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Documents
10108-450.pdf
Fiche technique
Spécifications
- Taille:50 µg
- Western blot:Yes
- Cat. No.:PRSI29-517
- Clonalité:Polyclonal
- Formulaire:Lyophilized
- Immunogène:A synthetic peptide corresponding a region of human EIF4E3.
- Symbole de l'antigène:EIF4E3
- Hôte:Rabbit
- Conjugaison:Unconjugated
- Réactivité:Human|Rat|Mouse
- ELISA:Yes
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



