Order Entry
Belgium
ContactUsLinkComponent
Anti-CASP8 Rabbit Polyclonal Antibody
Anti-CASP8 Rabbit Polyclonal Antibody
Catalog # PRSI3475
Supplier:  ProSci Inc.
undefined
Anti-CASP8 Rabbit Polyclonal Antibody
Catalog # PRSI3475
Supplier:  ProSci Inc.

Specifications

  • Nom de l'antigène:
    Caspase 8
  • Symbole de l'antigène:
    CASP8
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Mouse
  • Western blot:
    Yes
  • Synonymes antigène:
    FLICE|caspase 8|MACH-beta-1/2/3/4 protein|FADD-homologous ICE/CED-3-like protease|MCH5|apoptotic protease Mch-5|FADD-like ICE|MACH-alpha-1/2/3 protein|CAP4|ALPS2B|apoptotic cysteine protease|Casp-8|MORT1-associated ced-3 homolog|caspase-8|apoptosis-related cysteine protease|ICE-like apoptotic protease 5|MACH
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogène:
    Caspase-8 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus human caspase-8 isoform E.
  • Tested applications:
    E, WB
  • Purification:
    Affinity chromatography purified via peptide column
  • Taille:
    100 µg
  • Cdt:
    100 µG

Specifications

About this item

Caspase-8 Antibody: Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. The apoptotic subfamily can be further divided into initiator caspases, which are activated in response to death signals, and executioner caspases, which are activated by the initiator caspases and are responsible for cleavage of cellular substrates that ultimately lead to cell death. Caspase-8 is an initiator caspase that was identified as a member of the Fas/APO-1 death-inducing signaling complex. The adaptor molecule FADD couples procaspase-8 to the Fas receptor death domain; subsequent oligomerization promotes procaspase-8 autoactivation. FLIP, a catalytically inactive caspase-8-like molecule inhibits these interactions and thus can inhibit apoptosis. This antibody will only detect isoform E of caspase-8.

Type:
Antigen: CASP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse