Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Chlorothiazide, Reference Standard

Chlorothiazide, USP reference standard, Vial, 200 mg
N° de catalogue 1121005.
Fournisseur:   USP
Chlorothiazide, Reference Standard
N° de catalogue 1121005.
Fournisseur:  USP
Numéro CAS:  
Chlorothiazide, Reference Standard

Chlorothiazide, USP reference standard, Vial, 200 mg

Numéro CAS:  
Prix
405,00 €
Connexion Pour la disponibilité
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    200 mg
  • Normes:
    USP reference standard
  • Désignation:
    Chlorothiazide
  • Cat. No.:
    1121005.
  • N ° CAS.:
    58-94-6
  • Formula:
    C₇H₆ClN₃O₄S₂
  • Désignation:
    Chlorothiazide

Spécifications