Handheld microscope, BMS Trekker
Handheld microscope, 10× - 35×
N° de catalogue BREU76520
Fournisseur: BREUKHOVEN
Aller à :
Product Details & Documents
The BMS Trekker is a high quality handheld microscope that enhances objects up to 35 times.
- Detachable eyepiece to be used as a 10× magnification latch
- Built-in white LED with daylight correction – efficient energy saving lamp
- Can be connected to any SLR camera
Suitable for outdoor and indoor use in technology, science, schools and leisure.
Spécifications
- Grossissement:10× - 35×
- Désignation:Handheld microscope
- Cat. No.:BREU76520
- Poids:215 g
- Éclairage:LED
- Exigence électrique:9 V PP battery