Thermal unit for tissue embedding system TES 99
MEDT02-9960-00
: MEDITE
- Poids:15 kg
- Désignation:Thermal unit for tissue embedding system TES 99
- l×p×h:360×570×300 mm
- Environmentally Preferable:
- Cdt:1
The thermal unit features a prewarming chamber for embedding molds in the upper section. The lower section contains the working plate, and a removable paraffin basin made of stainless steel.
It is deep enough for storage of embedding cassettes and baskets. The stainless steel cover allows for temperature stability.