Handle for craft tool
Handle for craft tool
Handle, orange PS, without blades, 104 mm
N° de catalogue SWAN1235
Fournisseur:   SWANN MORTON
Handle for craft tool
N° de catalogue SWAN1235
Fournisseur:  SWANN MORTON
Handle for craft tool
Handle for craft tool
Handle, orange PS, without blades, 104 mm
Prix
44,30 €
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Handle made from impact resistant PS.
  • A thumbscrew is used for blade changing and retaining
  • For use with blades No. 1, 2 or 3
This craft tool is an ideal tool for marquetry, modelling, craft, studio work and horticulture.
Ordering information: Handle only. Blades need to be ordered separately.
Spécifications
  • Désignation:
    Handle, orange PS, without blades
  • Longueur:
    104 mm