Equilibrium DIALYZER™ plate for Single Use 96-Well Equilibrium DIALYZER™
N° de catalogue HARV74-2331
Fournisseur: HARVARD APPARATUS
Spécifications
- Désignation:96-Well Equilibrium DIALYZER™ plate with 10 KDa MWCO membrane
- Cdt:1
Spécifications
À propos de cet article
The Single Use 96-Well Equilibrium DIALYZER™ is designed for high throughput protein interaction studies.
- Each well in this system has a separate membrane, eliminating possibility of cross-contamination
- Excellent sample recovery
- Wells are sealed with 8-cap strips (not included)