TM Buffer, 1X Tris-Magnesium sulphate, pH 7,5
Catalog # ROCKMB-059
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Cdt:1 L
- Désignation:TM buffer (50 mM tris Hcl, pH 7,5, 10 mM magnesium sulfate)
- Seq:16
Specifications
About this item
This product is ready-to-use as a working 1X solution and requires no further dilution. This buffer consists of 50 mM Tris HCl, 10 mM magnesium sulphate at pH 7,5 and was prepared using ultra pure reagents dissolved in pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.