TES buffer, 1X, ready to use, pH 7,8
Catalog # ROCKMB-058
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Cdt:1 L
- Désignation:TES 1X , pH 7,8 (10 mM tris Hcl, 100 mM Nacl, 1 mM EDTA) DEPC treated
- Seq:15
Specifications
About this item
This product is ready-to-use as a working 1X solution and requires no further dilution. This buffer consists of 10 mM Tris HCl, 100 mM NaCl, and 1,0 mM EDTA at a pH of 7,8. Meticulously prepared using ultra pure reagents dissolved in DEPC treated pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.