Sample preparation kit, AdvanceBio
N° de catalogue HEWL5190-8002
Fournisseur: Agilent
Spécifications
- Désignation:AdvanceBio n-glycan deglycosylation cleanup cartridges, 24 samples
- Cdt:1
Spécifications
À propos de cet article
This kit consist the enzymes and cleanup cartridges for the enzymatic deglycosylation process of N-glycan.
Deglycosylation kit includes PNGase F enzyme, phosphate reaction buffer, TRIS reaction buffer, detergent solution, denaturation solution.
Labelling kit includes 75 µl 2-aminobenzeamide solution and 75 µl reductant (sodium cyanoborohydride) solution.
Livraison: Deglycosylation kit, labelling kit and cleanup cartridges.