Anti-EIF4G1 Rabbit Polyclonal Antibody (Cy7®)
The protein encoded by this gene is a component of the multi-subunit protein complex EIF4F. This complex facilitates the recruitment of mRNA to the ribosome, which is a rate-limiting step during the initiation phase of protein synthesis. The recognition of the mRNA cap and the ATP-dependent unwinding of 5'-terminal secondary structure is catalyzed by factors in this complex. The subunit encoded by this gene is a large scaffolding protein that contains binding sites for other members of the EIF4F complex. A domain at its N-terminus can also interact with the poly(A)-binding protein, which may mediate the circularization of mRNA during translation. Alternative splicing results in multiple transcript variants, some of which are derived from alternative promoter usage. [provided by RefSeq].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EIF4G1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Adsorption croisée:No
- Type d'anticorps:Primaire
- Formulaire:liquid
- Purification:Purified by Protein A
- Symbole de l'antigène:EIF4G1
- Synonymes antigène:EIF-4G1|PARK18|EIF4GI|EIF4F|EIF4G|P220
- Isotype:IgG
- Conjugaison:Cy7®
- Concentration:1 μg/μl
- Réactivité:Human|Rat|Mouse
- Température de stockage:Store at 4°C for 12 months
- ID de gène:1981
- Cat. No.:BOSSBS-3843R-CY7
- Clonalité:Polyclonal
- Nom de l'antigène:eukaryotic translation initiation factor 4 gamma, 1
- Hôte:Rabbit
- ImmunoChimie:Yes
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de transport:4°C



