Anti-THEMIS Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-12329R-CY3
Supplier: Bioss
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Thymocyte-expressed Molecule Involved in Selection
- Symbole de l'antigène:THEMIS
- Clonalité:Polyclonal
- Conjugaison:Cy3®
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Environmentally Preferable:
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:387357
- Synonymes antigène:thymocyte-expressed molecule involved in selection|THEMIS|bA325O24.3|FLJ40584|thymocyte selection associated|bA325O24.4|TSEPA|thymocyte selection pathway associated|C6orf190|MGC163388|C6orf207
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Immunogène:300-350/641
- Purification:Purified by Protein A
- Cdt:100 µl
Specifications
About this item
Plays a central role in late thymocyte development by controlling both positive and negative T-cell selection. Required to sustain and/or integrate signals required for proper lineage commitment and maturation of T-cells. Regulates T-cell development through T-cell antigen receptor (TCR) signaling and in particular through the regulation of calcium influx and phosphorylation of Erk (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: THEMIS
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat