Anti-p-ZAP70 Rabbit Polyclonal Antibody [clone: Y315]
Catalog # BWRLBS4884
Supplier: Bioworld Technology
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:p-ZAP70
- Clonalité:Polyclonal
- Clone:Y315
- Hôte:Rabbit
- ImmunoChimie:Yes
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human ZAP70 around the phosphorylation site of Tyrosine 315.
p-ZAP70 (Y315) pAb detects endogenous levels of ZAP70 protein only when phosphorylated at Tyr315.
Type: Primary
Antigen: p-ZAP70
Clonality: Polyclonal
Clone: Y315
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat