Order Entry
Belgium
ContactUsLinkComponent
 
Anti-p-PKD1 Rabbit Polyclonal Antibody [clone: S910]
Catalog # BWRLBS4862
Anti-p-PKD1 Rabbit Polyclonal Antibody [clone: S910]
Catalog # BWRLBS4862

Specifications

  • Type d'anticorps:
    Primaire
  • Symbole de l'antigène:
    p-PKD1
  • Clonalité:
    Polyclonal
  • Clone:
    S910
  • Hôte:
    Rabbit
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Taille:
    100 µg
  • Cdt:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human PKD1 around the phosphorylation site of Serine 910.

p-PKD1 (S910) pAb detects endogenous levels of PKD1 protein only when phosphorylated at Ser910.

Type: Primary
Antigen: p-PKD1
Clonality: Polyclonal
Clone: S910
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat