Anti-p-Synapsin I Rabbit Polyclonal Antibody [clone: S9]
Catalog # BWRLBS4802
Supplier: Bioworld Technology
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:p-Synapsin I
- Clonalité:Polyclonal
- Clone:S9
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Synapsin I around the phosphorylation site of Serine 9.
p-Synapsin I (S9) pAb detects endogenous levels of Synapsin I protein when phosphorylated at Ser9.
Type: Primary
Antigen: p-Synapsin I
Clonality: Polyclonal
Clone: S9
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat