Benchtop meter-attached electrode stand, Orion™ Star A series
N° de catalogue ORIOSTARA-BEA
Fournisseur: Thermo Orion
Spécifications
- Pour:Appareils de mesure pour paillasse Orion Star A110 + A210, Orion Versa Star
- Désignation:Support d'électrode relié à un appareil de mesure pour paillasse (bras et tête de support d'électrode), Orion™ Star A, mouvement à 360° (sans base lestée)
- Seq:103
- Environmentally Preferable:
- Cdt:1
Spécifications
À propos de cet article
This meter-attached electrode stand can be used to easily place sensors in samples.
- Simplifies rinsing and storage between samples
- Cable management system minimises electrode breakage
- 360° motion allows electrodes to be moved vertically, horizontally and front-to-back positioning into beakers or sample containers
Can be attached to either side of Thermo Scientific™ Orion™ Versa Star benchtop meters and Thermo Scientific™ Orion™ Star A210 and Star A110 series benchtop meters. Can also be used with weighted base for compatibility with any meter system (portable or benchtop).
Informations de commande: The weighted base needs to be ordered separately.
Livraison: Includes meter bracket, arm and electrode holder.