Distillation apparatus
Catalog # NEUB9100D1160-0002
Supplier: NEUBERT VOLUME GLASSWAERE
Specifications
- Désignation:Distillation apparatus complete with thermometer, tripod and heating hood
- Cdt:1
Specifications
About this item
Reduced Pressure distillation apparatus according to ASTM D 1160.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
5-piece made of glass with additional 3×ball-joint clamp complete with KS 35 and NS19/38. Cylinder double wall 200 ml graduated.