Cutlery, multi-purpose, Corning® Gosselin™
GOSSCFM-01
: Corning
- Longueur:180 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:RECs, ISO 14001 - Borre
- Cdt:500
Multi-purpose cutlery made from white PS. For sample collection and processing in food, pharmaceutical or cosmetic industries.
- Food contact approved: Raw material is compliant with regulation EC N°1935/2004
- Sterility assurance level based on ISO 11137: STERILE SAL 10⁻³
: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904