HPLC performance kit
N° de catalogue REST26394
Fournisseur: Restek
Spécifications
- Désignation:Performance kit for HPLC
- Cdt:1
Spécifications
À propos de cet article
This HPLC PEEK performance kit is a useful complement for the HPLC system.
Livraison: It includes 1 hub-cap adapter w/cap, 3 tubing clips, 1 clean cut tubing cutter, 5 PEEK finger tight fittings, 5 PEEK column plugs, 2 PEEK 90° tubing elbows, 2 PEEK 180° tubing elbows and 2 PEEK column connectors.