FID maintenance kits
Catalog # REST22179
Supplier: Restek
Specifications
- Désignation:FID maintenance kit
- Cdt:1 KIT
Specifications
About this item
FID maintenance kits for use with Agilent 6850, 6890, and 7890 GCs.
FID maintenance kits for use with Agilent 6850, 6890, and 7890 GCs.