Micropacked inlet conversion kits
Micropacked inlet conversion kits
Injection port adapter kit
N° de catalogue REST22426
Fournisseur:   Restek
Micropacked inlet conversion kits
N° de catalogue REST22426
Fournisseur:  Restek
Micropacked inlet conversion kits
Micropacked inlet conversion kits
Injection port adapter kit
Prix
144,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Micropacked inlet conversion kits are used to convert a capillary GC split/splitless inlet for use with ¹/₁₆" Ext.Ø micropacked columns.
  • For use with Agilent 5890 6890 and 7890 GCs
  • Sample pathways deactivated for ultimate inertness
Livraison: Kit includes dual large bore Vespel ring inlet seal, large bore reducing nut, large bore FID adaptor, ¹/₁₆" Vespel/graphite ferrule, ¹/₁₆" stainless steel nut, 4 mm splitless liner (intermediate polarity deactivated).
Spécifications
  • Désignation:
    Injection port adapter kit