Injector wrench for Shimadzu
Catalog # REST21159
Supplier: Restek
Specifications
- Désignation:Injector wrench
- Matière:Acier inoxydable
- Cdt:1
Specifications
About this item
This stainless steel inlet wrench is ideal for Shimadzu 17A, 2010, and 2014 GCs.
- Designed specifically for removing Shimadzu injection ports