Capillary columns, MXT®-1HT
Capillary columns, MXT®-1HT
 :   Restek
Capillary columns, MXT®-1HT
 :  Restek
Capillary columns, MXT®-1HT
Capillary columns, MXT®-1HT
MXT®-1HT Siltek®-treated stainless steel nonpolar phase columns.
  • Lowest bleed for longest column lifetime
  • 100% dimethyl polysiloxane phase allows easy comparisons to historical data
  • Individually tested
  • 177,8 mm coil diameter
  • Temperature ranges from 60 to 450 °C
Certifications: Meets all ASTM D2887, D6352, D7169, D7213, and D7500 specifications.
Product Family Options
  • Type de colonne
    Greffage
    Épaisseur du film
    Ø int.
    Longueur
  • Capillaire
    MXT®-1HT
    0,10 µm
    0,53 mm
    5 m
    1 * 1 ST
    1 * 1 ST - 570,00 €
    570,00 €
    1 * 1 ST
       570,00 €
  • Capillaire
    MXT®-1HT
    0,20 µm
    0,53 mm
    5 m
    1 * 1 ST
    1 * 1 ST - 570,00 €
    570,00 €
    1 * 1 ST
       570,00 €
  • Capillaire
    MXT®-1HT
    0,88 µm
    0,53 mm
    5 m
    1 * 1 ST
    1 * 1 ST - 740,00 €
    740,00 €
    1 * 1 ST
       740,00 €
  • Capillaire
    MXT®-1HT
    0,21 µm
    0,53 mm
    10 m
    1 * 1 ST
    1 * 1 ST - 775,00 €
    775,00 €
    1 * 1 ST
       775,00 €
  • Capillaire
    MXT®-1HT
    0,88 µm
    0,53 mm
    10 m
    1 * 1 ST
    1 * 1 ST - 775,00 €
    775,00 €
    1 * 1 ST
       775,00 €
  • Capillaire
    MXT®-1HT
    1,00 µm
    0,53 mm
    10 m
    1 * 1 ST
    1 * 1 ST - 775,00 €
    775,00 €
    1 * 1 ST
       775,00 €
  • Capillaire
    MXT®-1HT
    1,20 µm
    0,53 mm
    10 m
    1 * 1 ST
    1 * 1 ST - 775,00 €
    775,00 €
    1 * 1 ST
       775,00 €
  • Capillaire
    MXT®-1HT
    2,65 µm
    0,53 mm
    10 m
    1 * 1 ST
    1 * 1 ST - 871,00 €
    871,00 €
    1 * 1 ST
       871,00 €
  • Capillaire
    MXT®-1HT
    5,00 µm
    0,53 mm
    10 m
    1 * 1 ST
    1 * 1 ST - 871,00 €
    871,00 €
    1 * 1 ST
       871,00 €