ContactUsLinkComponent
In-situ hybridisation buffer for HPV probes
In-situ hybridisation buffer for HPV probes
Catalog # ENZOENZ339050050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
In-situ hybridisation buffer for HPV probes
Catalog # ENZOENZ339050050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Cdt:
    50 mL
  • Désignation:
    In situ hybridisation buffer for HPV probes
  • Seq:
    02
Specifications
About this item
In situ hybridisation buffer for HPV probes (ready-to use) provides a high quality hybridisation medium for In situ hybridisation analyses using either DNA or oligonucleotide probes labeled with biotin-, or fluorescein-modified nucleotides.
  • Optimised buffer to use with HPV collection of probes