Vu-tight installation fittings kit
Catalog # REST20504
Supplier: Restek
Specifications
- Désignation:Vu-tight installation fittings kit
- Cdt:1
Specifications
About this item
This kit includes a ¹/₄" stainless steel nut and graphite ferrule for attaching the liner to the GC inlet and a ¹/₄ to ¹/₁₆" stainless steel reducer, plus a ¹/₄" and 0,5 mm ID graphite ferrule for attaching the column to the liner.